Cryogenic Liquid Labels, Labelmaster
Supplier: Label Master
L31R
101376-246PK 189.07 USD
101376-246
Cryogenic Liquid Labels, Labelmaster
Labels
Comply with international air regulations by using cryogenic liquid labels.
  • Must be used with a non-flammable gas label when shipping cryogenic materials by air
  • Printed on paper
  • Outstanding chemical and abrasion resistance
  • Green and white
Labels help to shipments in regulatory compliance.