Elite-Q PLOT Phase Columns, PerkinElmer
Supplier: PerkinElmer
N9316360
10137-180EA 2582.99 USD
10137-180
Elite-Q PLOT Phase Columns, PerkinElmer
Chromatography Columns
Elite-Q PLOT Phase Columns are for analysis of light gases and hydrocarbons.
  • Available in a range of inner diameter measurements
  • Column length is 30 meters
  • Suited for analysis of light cases and hydrocarbons
PLOT columns are the best choice for analysis of highly volatile compounds such as permanent gases, solvents, and volatile petrochemicals such as refinery gases.