Stainless Steel ATD Prepacked Sample Tubes, PerkinElmer
Supplier: PerkinElmer
N9307002
10136-870PK 3298.34 USD
10136-870
Stainless Steel ATD Prepacked Sample Tubes, PerkinElmer
Tubes Test Tubes
These prepacked sample tubes are designed for Carbopack™ B 60/80.
  • Analyte volatility range is n-C5 to n-C12
The Carbopack™ B is conditioned at 350 °C. These tubes are deigned with maximum temperature of 400 °C with a desorption temperature of 330 °C.