Snap Top Cap with PTFE/Silicone, PerkinElmer
Supplier: PerkinElmer
N9303416 N9303419
10136-820PK 86.22 USD
10136-820 10139-724
Snap Top Cap with PTFE/Silicone, PerkinElmer
Closures Chromatography Vial Closures
Snap top closures are designed in a wide variety of sizes and materials based on the sample type and analysis temperature
  • Contaminant free product
  • Excellent resealability
  • Multiple injection capability
Closures are made with specific tolerances that are guaranteed to fit and perform with all type of instruments.
Using a process that bonds silicone/PTFE and otherelastomeric compounds directly into thermoplastic closures eliminates liner fallout.