MicroFlow™ PFA Self-Aspirating Nebulizers, PerkinElmer
Supplier: PerkinElmer
N8145102
10136-716EA 3734.21 USD
10136-716
MicroFlow™ PFA Self-Aspirating Nebulizers, PerkinElmer
Atomizers
An integrated capillary PFA nebulizer for self aspiration.
  • Resistant to clogging
  • Produces a fine aerosol for high transport efficiency and high sensitivity
  • Low, spike free background for important elements such as Fe and Ca
  • Direct analysis of volatile and non-volatile organic solvents
PFA MicroFlow nebulizers are constructed entirely from chemically resistant fluoropolymers, ideal for strong acids, alkalis, organics and high salt solutions.