MEINHARD™ Nebulizer Clip Snapper for Avio® 200/500 and Optima™ 2×00/4×00/5×00/7×00 DV/8×00, PerkinElmer
Supplier: PerkinElmer
N0777460
10136-464EA 21.94 USD
10136-464
MEINHARD™ Nebulizer Clip Snapper for Avio® 200/500 and Optima™ 2×00/4×00/5×00/7×00 DV/8×00, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
This nebulizer clip snapper, for MEINHARD™ nebulizers, is used with Optima™ 4×00/5×00/7×00/8×00 and Avio® 200/500 ICP-OES series instruments in addition to ELAN® 5000/6×00/9000/DRCs and NexION® 300/350/1000/2000 ICP-MS series instruments.
This device is included with all standard, plus, and SilQ MEINHARD™ nebulizers.