Demountable Quartz Injector, PerkinElmer
Supplier: PerkinElmer
N0777457
10136-462EA 2482.66 USD
10136-462
Demountable Quartz Injector, PerkinElmer
Chromatography Supplies Chromatography Injectors
This Quartz Injector Cassette Mount is designed for use with demountable torches in PerkinElmer NexION® 300/350 ICP-MS series instruments.
  • 2.0 mm inner diameter
  • Aassembly base is made from PFA and includes O-Rings
The demountable design allows the injector to be removed and replaced efficiently without dismounting the torch.