Sapphire Injector for Demountable ZipTorch for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Supplier: PerkinElmer
N0777044
10136-390EA 4414.95 USD
10136-390
Sapphire Injector for Demountable ZipTorch for PerkinElmer Optima ICP-OES Instruments, PerkinElmer
Microinjectors Microinjectors, Programmable
This demountable injector is for use with Optima® 4x00/5x00/7x00 ICP-OES series instruments.
It is designed for use with the quartz, organics and silicon nitride demountable ZipTorches. The demountable design allows the injector to be removed and replaced efficiently for cleaning without dismounting the torch.