Replacement CTFE Stator for High Flow Valve, PerkinElmer
Supplier: PerkinElmer
N0771651
10136-378EA 6903.9 USD
10136-378
Replacement CTFE Stator for High Flow Valve, PerkinElmer
Chromatography Supplies Chromatography Spare Parts
High-purity valve stators and rotors provide a liquid path with low contamination and low carryover for the best ICPMS results.
  • CTFE stators offers high impact strength, chemical, and corrosion resistance
  • Wide operational temperature range: –76 to 150 °C (–105 to 300 °F)
This replacement stator features six ports. The bore size is 0.8 mm.