Complete Quartz Torch O-Ring Kit for Optima 4300/5300/7300 V, PerkinElmer
Supplier: PerkinElmer
N0770916
10136-370EA 176.2 USD
10136-370
Complete Quartz Torch O-Ring Kit for Optima 4300/5300/7300 V, PerkinElmer
Seals Sealing Rings
Various torch assembly O-rings used with Optima™ 4300 V, 5300 V, and 7300 V ICP-OES series instruments are packaged in a single kit.
This kit includes: O-ring Int. Ø 15.5 mm, Qty.1 (97012-114), O-ring Int. Ø 6.07 mm, Qty.2 (97012-126), O-ring Int. Ø 9.25 mm, Qty.2 (97012-130), O-ring Int. Ø 7.65 mm, Qty.1.