Rectangular Zinc Selenide Sealed Cell, PerkinElmer
Supplier: PerkinElmer
L1270970
10136-366EA 4933.08 USD
10136-366
Rectangular Zinc Selenide Sealed Cell, PerkinElmer
Cuvettes Spectrophotometer Cuvettes
Sealed liquid cells are designed as a preassembled integral unit. They are ideal for highly volatile samples and quantitative analysis.
  • Made of zinc selenide
  • Pathlength of 0.1 mm
The sealed liquid cell has a corrosion-resistant construction and a precise pathlength which is calibrated using the interference pattern generated by an empty cell in the beam of a spectrometer.