KRS-5 Circular Demountable Cell Windows, PerkinElmer
Supplier: PerkinElmer
L1270665
10136-362EA 3212.05 USD
10136-362
KRS-5 Circular Demountable Cell Windows, PerkinElmer
Cuvettes Spectrophotometer Cuvettes
Circular Demountable Cell Windows are available in a variety of window materials, including Thallium Bromide-Iodide (KRS-5).
  • Thickness: 4 mm
The windows fit circular cells and have a diameter of 25 mm.