Flow Injection System Supplies, PerkinElmer
Supplier: PerkinElmer
B0508306 B0507962 B0507957 B0507959 B0187258 B0193772
97012-768PK 1158.28 USD
97012-768 97013-224 10136-346 10136-348 10139-018 10139-026
Flow Injection System Supplies, PerkinElmer
Flow Meters
Several of modular building block type mixing manifold blocks can be connected in series for enhanced mixing capabilities.
  • Modular building blocks are made of chemically-resistant plastic
Modular building block type gas/liquid separator: An exchangeable PTFE membrane in the screw cap prevents liquid from being carried into the quartz cell when working with strong foaming samples.
Delivery information: Complete modular mixing/separation assembly includes two mixing manifolds with tubing adapters, gas liquid separator with PTFE membrane, 5 spare PTFE membranes, PTFE tubes (110 and 300 mm). Modular building block type mixing manifold includes mixing channel and 3 connections, 2 inlet and 1 outlet.