Autosampler Sample Pans, 30 µl, PerkinElmer
Supplier: PERKINELMER U.S. LLC
B0169320
10136-334EA 3045.61 USD
10136-334
Autosampler Sample Pans, 30 µl, PerkinElmer
Trays/Dishes/Pots Laboratory Trays/Dishes
The Autosampler sample pans are used to prepare sample materials for PerkinElmer DSC Autosampler. These pans have an operating range of –170 to 600 ° C, a volume of 30 µl, and 3 bar maximum pressure.
  • Used by the universal crimper press (10137-514)
  • Can be used with all autosamplers
These sample pans are for use with Diamond DSC, Pyris 1 DSC, DSC 7, DSC 6, Pyris 6 DSC, and Jade DSC. These pans are hermetically sealed and may also be used for non-robotic systems.
Delivery information: Includes 400 sample pans.