EzyLok Nebulizer Pressure Hose Clips, PerkinElmer
Supplier: PerkinElmer
N0773197
10136-330PK 107.12 USD
10136-330
EzyLok Nebulizer Pressure Hose Clips, PerkinElmer
Clamps Tubing and Sanitary Clamps
Hose clips are designed for use with Avio® 200/500 and Optima™ 4×00/5×00/7×00/8×00 ICP-OES series instruments.
  • Designed for use with EzyLok tubing in SeaSpray™ and MicroMist™ nebulizers
These clips are included in the EzyLok kit for SeaSpray™ and MicroMist™ nebulizers (10139-382).