Demountable Quartz Torch, 3-Slot, for Optima 2x00/4x00/5x00/7x00 DV, PerkinElmer
Supplier: PerkinElmer
N0772005
10136-320EA 1097.99 USD
10136-320
Demountable Quartz Torch, 3-Slot, for Optima 2x00/4x00/5x00/7x00 DV, PerkinElmer
Lights Flashlights
This demountable 3-slot torch design uses one-piece quartz tubing for plasma and auxiliary gas flows. This style torch is recommended for use in organic analysis.
  • Demountable design permits quick changing of the torch body only, saving time and money
  • Recommended for use in organic analyses
This 3-Slot quartz torch is compatible with the Optima™ 2x00 DV, 4x00 DV, 5x00 DV, and 7x00 DV ICP-OES series PerkinElmer instruments.