Torch Module Support Stand, PerkinElmer
Supplier: PerkinElmer
N0770609
10136-312EA 618.83 USD
10136-312
Torch Module Support Stand, PerkinElmer
Lights Flashlights
The free-standing support stand is used with Optima™ 2x00 DV, 4x00 DV, 5x00 DV, 7x00 DV, and 8x00 ICP-OES series instruments.
A free-standing support stand to hold spare quick-change torch module or as a means of conveniently holding module when working inside the sample compartment.