Barium Fluoride Semi-Demountable Cell Window, PerkinElmer
Supplier: PerkinElmer
L1271194
10136-268EA 3013.37 USD
10136-268
Barium Fluoride Semi-Demountable Cell Window, PerkinElmer
Cuvettes Spectrophotometer Cuvettes
Barium Fluoride (BaF₂) is a relatively hard material, similar to CaF₂.
  • Windows are polished to precise tolerances of flatness
  • Thickness: 4 mm
  • One pair
PerkinElmer offers a versatile product line of liquid sampling cells which utilize a specially designed universal mount suitable for both FT-IR and dispersive instruments, and a wide variety of high-quality window materials to meet varied analytical needs. The first criteria to consider when selecting a window material is the wavelength range over which the spectrum must be measured. Other important considerations include solubility, reactivity, and the refractive index of the window material with respect to the sample. Sometimes mechanical and thermal characteristics may also be important.