Zinc Selenide Semi-Demountable Cell Windows, PerkinElmer
Supplier: PerkinElmer
L1270765
10136-238EA 3306.89 USD
10136-238
Zinc Selenide Semi-Demountable Cell Windows, PerkinElmer
Specimen Mounting Supplies
Zinc Selenide (Irtran-4™) is similar to Cleartran except that it is somewhat softer and more easily scratched.
  • Specially designed universal mount suitable for both FT-IR and dispersive instruments
  • Polished to precise tolerances of flatness
Irtran-4 is used in total reflectance accessories because of its high refractive index.