HGA Graphite Contact Cylinders, 5 Pairs, PerkinElmer
Supplier: PERKINELMER U.S. LLC
B3130086
10136-184EA 1724.11 USD
10136-184
HGA Graphite Contact Cylinders, 5 Pairs, PerkinElmer
Tubes Graphite Tubes
The graphite contact cylinders are engineered for a precise fit so that variations in electrical contact, which might affect the analytical performance, are reduced to a minimum.
PerkinElmer has over four decades of experience in the design/development of graphite furnace systems and materials. The proper performance, maintenance and serviceability of our graphite furnace systems can be assured only through the use of genuine , PerkinElmer supplied graphite components such as these contact cylinders.
Fits PerkinElmer PinAAcle 900H, AAnalyst 700, HGA-900, HGA-800, HGA-700, HGA-600, HGA-300 AA instruments.
Delivery information: Each set includes one left-hand contact, one right-hand contact and one shielding ring.