Sampling Probes Assemblies for Autosamplers, PerkinElmer
Supplier: PerkinElmer
B3001770 B3001772 B3000159 B3000160 B3000152 B3000161 B3001771 N0777434 N8145139 B3001769
97013-182EA 787.71 USD
97013-182 97013-184 97013-192 97013-200 97013-208 97013-190 10136-176 10136-420 10136-728 10137-644
Sampling Probes Assemblies for Autosamplers, PerkinElmer
Electrodes Other Electrodes
Sampling probe assemblies for AS-90/90A/90plus/91/93plus/S10 autosampler.
Corrosion-resistant probes are suitable for inorganic acids and most organic solvents, except NMP.
PTFE probes provide laminar flow and are the most resistant to chemical degradation.
Delivery information: Corrosion-resistant sampling probe assemblies includes sampling Probe with FEP tube, screw fitting and capillary tubing assembly.