Polarimeter Cells, PerkinElmer
Supplier: PerkinElmer
B0017054
97012-496EA 1756.76 USD
97012-496 10136-166
Polarimeter Cells, PerkinElmer
Cuvettes Specialty Cuvettes
Cells are designed for easy flling and emptying.
  • High efficiency and thermostattable
  • Filling ports are arranged so that the liquid flows over the inner window surfaces
  • Windows are fused to the cell bodies to ensure tension-free operation
  • Defined pathlength engraved on each cell for easy identification
  • Cells are jacketed for thermostatting with constant temperature water, except the flowcell
Use quartz cells for all wavelengths and for elevated temperatures (200 °C, maximum). Use glass cells only for visible wavelengths, above 400 nm.
Cells are individually tested with respect to its pathlength. The actual pathlength of each cell is engraved on
the cell to the nearest 0.001 cm, together with the serial number of the cell.