HardyDisks™ Lysostaphin
Supplier: HARDY DIAGNOSTICS
Z112
10136-070EA 146.42 USD
10136-070
HardyDisks™ Lysostaphin
Microbiology Test Systems Antibiotic Tests
Differentiates between Micrococcus and Staphylococcus.
Used to rapidly differentiate Staphylococcus spp. and Micrococcus spp. based on lysostaphin resistance. Make a suspension of the bacteria, add the disk, look for a clearing of turbidity if Staph is present.