BioArray™ 5-hmC Methylation Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-45011-0050
10136-008EA 810.96 USD
10136-008
BioArray™ 5-hmC Methylation Kit, Enzo Life Sciences
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Detect 5-hydroxymethylcytosine using a robust glucosylation method
The BioArray™ 5-hmC methylation kit allows for sequence specific detection of 5-hydroxymethylcytosine within DNA using a simple and efficient glucosylation reaction procedure.
Ordering information: Contains 5-hmC Glucosyltransferase (5-hmC), 10X 5-hmC Reaction buffer, 10X UDPG (uridine diphosphoglucose), MspI restriction enzyme, 5-hmC Control DNA, qPCR Primer 1, qPCR Primer 2 and 5-DNA clean up concentrator kit .