Express DNA Methylation Kit, Deep Well, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-45002-0002
10136-000EA 794.61 USD
10136-000
Express DNA Methylation Kit, Deep Well, Enzo Life Sciences
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
Fastest method for complete, high-throughput (96-well) bisulfite conversion of DNA for methylation analysis.
The Express DNA methylation kit (deep well) features high-throughput (96-well) bisulfite treatment and conversion of DNA for methylation analysis. No preparation is necessary when using the Express Conversion Reagent. Simply add this reagent to a DNA sample and let the reaction proceed to completion. DNA denaturation and bisulfite conversion processes are combined with added heat to facilitate rapid denaturation. Desulphonation and clean-up of the converted DNA is performed using a unique 96-well spin-plate.High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing.
Ordering information: Contains Express conversion reagent, Methylation binding buffer, Methylation wash buffer concentrate, L-Desulphonation reagent, Methylation elution buffer, Express binding plate (deep), Express conversion plates, Collection plates, Elution plates.