EpiXtract® Nuclear Protein Isolation Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-45016-0100
10135-966EA 309.56 USD
10135-966
EpiXtract® Nuclear Protein Isolation Kit, Enzo Life Sciences
Protein Extraction Protein Extraction Kits
Rapid, reliable method to isolate nuclear proteins in mammalian cells and tissues.
For the extraction of nuclear proteins from mammalian cells or tissues.
EpiXtract® Nuclear Protein Isolation Kit II provides a simple and reliable method to isolate nuclear proteins. Extracts can be used for a variety of downstream applications including western blotting, protein-DNA binding assays, nuclear enzyme assays and others requiring nuclear proteins.
Ordering information: Contains Pre-extraction Buffer, Extraction Buffer, Protease Inhibitor Cocktail, DTT.