EpiXtract® Nuclear Protein Isolation Kit II (nucleic acid-free), Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-45015-0100
10135-964EA 295.39 USD
10135-964
EpiXtract® Nuclear Protein Isolation Kit II (nucleic acid-free), Enzo Life Sciences
Protein Extraction Protein Extraction Kits
Rapid, reliable method to isolate nucleic acid-free nuclear proteins in mammalian cells and tissues.
For the extraction of nucleic acid-free nuclear proteins from mammalian cells or tissues.
EpiXtract® Nuclear Protein Isolation Kit II provides a simple and reliable method to isolate nucleic acid-free nuclear proteins. Extracts can be used for a variety of downstream applications including western blotting, protein-DNA binding assays, nuclear enzyme assays and others requiring nucleic acid-free nuclear proteins.
Ordering information: Contains Pre-extraction Buffer, Extraction Buffer, Extraction pre-cleaner, Extraction cleaner, Protease Inhibitor Cocktail, DTT.