EpiXtract® Total Histone Extraction Kit, Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-45014-0100
10135-962EA 250.7 USD
10135-962
EpiXtract® Total Histone Extraction Kit, Enzo Life Sciences
Protein Extraction Protein Extraction Kits
Rapid, reliable method to isolate histone proteins in mammalian cells and tissues for histone modification applications.
For the extraction of total histone proteins from mammalian cells or tissues.
EpiXtract® Total Histone Extraction Kit provides a simple and reliable method to isolate histone proteins in 1 hour. Extracts can be used for a variety of downstream applications such as post-translational modification assays.
Ordering information: Contains 10x Pre-Lysis Buffer, Lysis Buffer, Balance Buffer, DTT .