EpiXtract® Total Histone Extraction Kit (High Throughput), Enzo Life Sciences
Supplier: Enzo Life Sciences
ENZ-45013-0002
10135-960EA 395.67 USD
10135-960
EpiXtract® Total Histone Extraction Kit (High Throughput), Enzo Life Sciences
Protein Extraction Protein Extraction Kits
Rapid, reliable, high throughput method to isolate histone proteins in mammalian cells for histone modification applications.
For the extraction of total histone proteins from mammalian cells in a 96 well plate format.
EpiXtract® Total Histone Extraction Kit (High Throughput) provides a simple and reliable method to isolate histone proteins in 1 hour. Extracts can be used for a variety of downstream applications such as post-translational modification assays.
Ordering information: Contains Pre-Lysis Buffer, Lysis Buffer, Balance Buffer, DTT .