MacConkey Broth, USP
Supplier: HARDY DIAGNOSTICS
U125
10135-862PK 208.85 USD
10135-862
MacConkey Broth, USP
Media for Microbiology Dehydrated Media for Microbiology
For the detection of gram-negative, lactose fermenting bacilli.
For the detection of gram-negative, lactose fermenting bacilli. Contains a durham tube for gas detection.