O-Ring-Free Organics Glass Twister Spray Chamber with Helix and Gas Port for Optima, PerkinElmer
Supplier: PERKINELMER U.S. LLC
N0777731
10134-610EA 3289.85 USD
10134-610
O-Ring-Free Organics Glass Twister Spray Chamber with Helix and Gas Port for Optima, PerkinElmer
Atomizers
This O-ring-free 50 ml glass Twister spray chamber, with integrated Helix™ and auxiliary gas port, is designed for organics applications for use with Optima® 2x00 DV, 4x00 DV, 5x00 DV, 7x00 DV, and 8x00 ICP-OES series instruments. It is recommended for use with Oil Analysis applications. It features a 4 mm baffle and Oxygen port.
  • Material: Borosilicate glass
  • Volume: 50 ml
The baffled, cyclonic Twister spray chamber has a central transfer tube, which acts as a secondary particle separator, helping separate larger aerosol particles from the sample. This reduces solvent load in the plasma without compromising detection limits. The position of the baffle is carefully controlled to ensure smooth draining and improved precision. The baffle is cut at a sharp angle so that a large cross-section of the spray chamber is sampled, ensuring representative sampling and reduced matrix effects.
Although this spray chamber isn't hydrofluoric acid-resistant, precision is considered excellent and purity is considered good. Low RSD's due to highly accurate construction Application uptake range 0.2 to 3.0 ml/min Produces smaller mean droplet size than standard spray chamber Positive stop for reproducible nebulizer insertion Convenient UniFit™ drain connector.