Ideal Micro Drill Kit, CellPoint Scientific
Supplier: Cell Point Scientific
67-1200A
101326-502EA 430.77 USD
101326-502
Ideal Micro Drill Kit, CellPoint Scientific
Tool Sets
The Ideal Micro-Drill™ is designed for research applications that require surgical burrs and trephines.
  • Light-weight design and Rechargeable
  • No special tools needed to change burrs
  • Effective balancing and handling
  • One year warranty
  • Hard plastic case with die-cut foam for convenient storage
The drill is constructed of light-weight aluminum alloy for balance and control and is powered by a rechargeable 6 V nickel cadmium battery. Under normal operating conditions the unit will function for 8 hours between recharging.