(-)-4,5-Bis[(2R,5R)-2,5-dimethylphospholanyl](1,2-dimethyl-1,2-dihydropyridazine-3,6-dione)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: [catASium® MNN(R)Rh]

45-0750-100MG
101324-772EA 277.42 USD
101324-772
(-)-4,5-Bis[(2R,5R)-2,5-dimethylphospholanyl](1,2-dimethyl-1,2-dihydropyridazine-3,6-dione)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate ≥95%
(-)-4,5-Bis[(2R,5R)-2,5-dimethylphospholanyl](1,2-dimethyl-1,2-dihydropyridazine-3,6-dione)(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

catASium

MDL Number: MFCD07782009
CAS Number: 908128-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR