1,2-Bis((2S,5S)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S,S)-i-Pr-DUPHOS-Rh, (-)-1,2-Bis((2S,5S)-2,5-di-i-propylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

45-0211-500MG 45-0211-100MG 45-0211-2G
101324-756EA 798.28 USD
101324-756 101324-754 101324-758
1,2-Bis((2S,5S)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
1,2-Bis((2S,5S)-2,5-diisopropylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate

BPE, DUPHOS

Formula: C₃₄H₅₆BF₄P₂Rh⁻
MDL Number: MFCD07369040
CAS Number: 1000393-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR