Polysiloxane thio-functionalised, macroporous (metal absorbing resin) (Deloxan® THP II)
Supplier: STREM CHEMICALS INC MS
Synonyms: Deloxan® (Thio-Functionalized Polysiloxane)
14-1870-50G 14-1870-10G 14-1870-250G
100197-378EA 653.91 USD
100197-378 101324-542 100197-376
Polysiloxane thio-functionalised, macroporous (metal absorbing resin) (Deloxan® THP II)
Polysiloxane thio-functionalised
Deloxan® THP II Macroporous, Thiourea-Functionalized Polysiloxane (Metal Absorbing Resin)
Color and Form: Abrasion-proof spheres; Water content: 50-70%
Physical Characteristics: surface area 300-450m2/g (BET), pore volume 2.5-3.5 ml/g, density 0.55-0.65 kg/L (wet form)
Sold in collaboration with Evonik for research purposes only.