Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Kobayashi's Reagent, NaBArF24

11-0590-250MG 11-0590-1G
101324-528EA 251.94 USD
101324-528 101324-530
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate ≥98%
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate
Formula: C₃₂H₁₂BF₂₄Na
MDL Number: MFCD00043323
CAS Number: 79060-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR