(S,S)-(+)-3,6-Octanediol ≥99%

Supplier: STREM CHEMICALS INC MS

08-2038-250MG 08-2038-1G
101324-522EA 200.98 USD
101324-522 101324-524
(S,S)-(+)-3,6-Octanediol ≥99%
(S,S)-(+)-3,6-Octanediol
Formula: C₈H₁₈O₂
Melting Pt: 49…51 °C
MDL Number: MFCD01631088
CAS Number: 136705-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR