(R,R)-(-)-3,6-Octanediol ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: (3R,6R)-Octane-3,6-diol, (R,R)-(-)-3,6-Dihydroxy octane, (3R,6R)-(-)-3,6-Octanediol

08-2037-1G 08-2037-250MG
101324-520EA 585.97 USD
101324-520 101324-518
(R,R)-(-)-3,6-Octanediol ≥99%
(R,R)-(-)-3,6-Octanediol
Formula: C₈H₁₈O₂
Melting Pt: 49…51 °C
MDL Number: MFCD01631087
CAS Number: 129619-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR