Dimethyl sulfoxide-[D₆] (99.9% D) for NMR spectroscopy

Supplier: 0000013444

DLM-10-5 DLM-10-10X0.75 DLM-10-10X0.6 DLM-10-25 DLM-10-50 DLM-10-1000 DLM-10-10X1 DLM-10-10 DLM-10-100
101322-304EA 89 USD
101322-304 101322-312 101322-294 101322-276 101322-296 101322-306 101322-284 101322-278 101322-286 103384-560
Dimethyl sulfoxide-[D₆] (99.9% D) for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
Boiling Pt: 189 °C (5 mmHg)
Melting Pt: 20.2 °C
Density: 1.19 g/cm³ (20 °C)
Flash Pt: 82 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002090
CAS Number: 2206-27-1
EINECS: 218-617-0

Order Now

Specification Test Results

Chemical Purity 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR