Dimethyl sulfoxide-[D₆] (99.96% D) for NMR spectroscopy

Supplier: 0000013444

DLM-34-10X0.75 DLM-34-5X0.5 DLM-34-10X0.5 DLM-34-5
101322-270EA 293.08 USD
101322-270 101322-272 101322-274 101322-266 101322-268 101322-262
Dimethyl sulfoxide-[D₆] (99.96% D) for NMR spectroscopy
Dimethyl sulphoxide-[D6]
Formula: D₃CS(O)CD₃
MDL Number: MFCD00002090
CAS Number: 2206-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR