Chloroform-[D₁] “100%” (D, 99.96%) +0.03% v/v TMS for NMR spectroscopy

Supplier: CAMBRIDGE ISOTOPE LABS INC MS
Danger

DLM-29-10X0.75
101322-172EA 91.15 USD
101322-172
Chloroform-[D₁] “100%” (D, 99.96%) +0.03% v/v TMS for NMR spectroscopy
Chloroform-[D1]

  • Contains 0.03% v/v TMS
  • No stabilizers are used in the product

Formula: CDCl₃
Density: 1.50
MDL Number: MFCD00000827
CAS Number: 865-49-6
UN: 1888

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR