Chloroform-[D₁] (99.8% D) +0.05% V/V TMS for NMR spectroscopy

Supplier: 0000013444
Warning

DLM-7TB-50 DLM-7TB-10X1 DLM-7TB-100
101322-156EA 54.92 USD
101322-156 101322-184 101322-154
Chloroform-[D₁] (99.8% D) +0.05% V/V TMS for NMR spectroscopy
Chloroform-[D1]

Contains 0.05% TMS.

Formula: CDCl₃
Boiling Pt: 60.8 °C (1013 hPa)
Melting Pt: –64.1 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000827
CAS Number: 865-49-6
EINECS: 212-742-4
UN: 1888
ADR: 6.1,III

Order Now

Specification Test Results

Purity 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR