Benzene-[D₆] (99.96% D) +0.03% (v/v) TMS, 99.5%

Supplier: 0000013444

DLM-40TC-10X0.75
101322-126EA 266.54 USD
101322-126
Benzene-[D₆] (99.96% D) +0.03% (v/v) TMS, 99.5%
Benzene-[D6]
Formula: C₆D₆
MDL Number: MFCD00003010
CAS Number: 1076-43-3
UN: 1114
ADR: 3,II

Order Now

Specification Test Results

Chemical Purity 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR