Calgonate® Hydrofluoric Acid Burn Relief Gel, Calgonate®
Supplier: Calgonate Corporation
CALGEL25
101320-858PK 214.29 USD
101320-858
Calgonate® Hydrofluoric Acid Burn Relief Gel, Calgonate®
First Aid Supplies Burn Care Supplies
Effective 2.5% topical calcium gluconate gel for hydrofluoric acid burn relief.
  • Provides instant pain relief and can be easily self-applied
Calgonate® gel is packaged in a 25 g (0.9 oz.) squeeze tube.