Decon 90, PerkinElmer
Supplier: PERKINELMER U.S. LLC
09923462
10132-302EA 104.12 USD
10132-302
Decon 90, PerkinElmer
Cleaning Agents Detergents and Surface Cleansers
Active surface cleaning agent, radioactive decontaminant for laboratory, medical and industrial applications.
  • Milky white, non-viscous liquid, having faint odour
  • A liquid concentrate, for dilution with water
  • Biodegradable, phosphate-free, Bactericidal, non-flammable and totally rinsable
  • For cleaning and decontaminating all manner of surfaces
  • Not suitable for use on non-ferrous metals, notably aluminium and zinc or polycarbonate
  • Problem contaminants can be treated by increasing the solution strength
Cleaning time can be reduced by heating the solution (40 to 50 °C) or using in an ultrasonic cleaning bath. The cleaned items must be rinsed thoroughly as soon as they are removed from the cleaning solution, and dried. We recommend three agitated rinses in water to ensure removal of all traces of contamination and cleaning agent.
Certifications: Ready biodegradability by OECD 301E (ISO method 7827 - 1986 (E)) and satisfies the criteria in BS EN 1040 1997 for bacterial activity.
Packaging: Supplied in 1 litre high density polyethylene container, with tamper-evident screw caps.