E.Z.N.A.® Blood DNA Kit, Omega Bio-tek
Supplier: OMEGA BIO-TEK
D3392-02
101319-436EA 20.57 USD
101319-436 101319-348 101319-412
E.Z.N.A.® Blood DNA Kit, Omega Bio-tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The E.Z.N.A.® Blood DNA Kit provides rapid total DNA isolation from 1–250 µL of fresh and frozen anticoagulated whole blood
This kit allows for simultaneous processing of single or multiple samples in less than 30 minutes. Phenol/chloroform extractions, and time-consuming steps such as precipitation with isopropanol or ethanol have been eliminated. DNA purified with the E.Z.N.A.® Blood DNA method is ready for applications such as PCR, Southern blotting, or restriction enzyme digestion.
The Kit can also be used for the preparation of genomic DNA from buffy coat, serum, plasma, bone marrow, lymphocytes, platelets, and body fluids.