E.Z.N.A.® Viral RNA Kit, Omega Bio-tek®
Supplier: OMEGA BIO-TEK
The E.Z.N.A.® Viral RNA Kit is designed for the isolation of viral RNA from cell-free fluids such as plasma, serum, urine, and cell culture supernatant
The procedure completely removes contaminants and enzyme inhibitors making viral RNA isolation fast, convenient, and reliable. The kit is also suitable for the isolation of total RNA from cultured cells, tissues, and bacteria. RNA purified using the E.Z.N.A.® Viral RNA method is ready for all downstream applications such as RT-PCR.