E.Z.N.A.® Plasmid Mini Kit I, Omega Bio-tek®
Supplier: OMEGA BIO-TEK
D6943-02 D6943-00 D6942-02 D6943-01 D6942-00 D6942-01
101318-898EA 241.34 USD
101318-898 101318-870 101318-896 101318-910 101318-868 101318-908
E.Z.N.A.® Plasmid Mini Kit I, Omega Bio-tek®
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The E.Z.N.A.® Plasmid Mini Kit I is designed to isolate up to 30µg of high-quality plasmid DNA from 1–5mL bacterial cultures in 30 minutes or less
Plasmid DNA purification is simplified with HiBind® Mini Column technology into three quick steps: Bind, Wash, and Elute. Purified plasmid DNA is immediately ready for a wide variety of downstream applications such as routine screening, restriction enzyme digestion, and manual/automated fluorescent DNA sequencing. E.Z.N.A.® Plasmid Mini Kit I is available in V- Spin and Q-Spin column formats.
Certifications: UL Listed.