Alumina Injectors for Optima ICP-OES Instruments, PerkinElmer
Supplier: PerkinElmer
N0775227 N0775228 N0775177 N0771531 N0582184 N0781007 N0781009 N0781014 N0781015
97012-838EA 859.08 USD
97012-838 97012-840 97012-842 97012-844 97012-832 10130-438 10130-440 10130-468 10130-470
Alumina Injectors for Optima ICP-OES Instruments, PerkinElmer
Microinjectors Microinjectors, Programmable
This Alumina Injector is used with Optima® 4300 V/5300 V/7300 V ICP-OES series instruments.
  • Alumina injectors are corrosion-resistant to all mineral acids including HF and aqua regia
  • Used with less volatile organic solvents, e.g., xylene
For injectors CA97012-838, CA97012-840, CA97012-842, 10130-440, 10130-468, 10130-470, the injector support adapter for Optima 2X00/4X00/5X00/7X00 DV 8x00 (CA97013-664) is compatible.
Delivery information: This Alumina Injector is included in the Quick-Change torch module for oil analysis and the organics sample introduction kit for Optima 4300 V/5300 V/7300 V (10136-316).