DSC Sample Pans, PerkinElmer
Supplier: PerkinElmer
N5390123
10130-462PK 592.15 USD
10130-462
DSC Sample Pans, PerkinElmer
Trays/Dishes/Pots Laboratory Trays/Dishes
Kit-Dsc sample pans, standard sample pans made up of aluminum.
Packaging: Pack of 100.